missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium Trichloride, TRC

Product Code. 30240668
Change view
Click to view available options
Quantity:
50 mg
250 mg
2.5 g
Unit Size:
2.5g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240668 2.5 g 2.5g
30240998 250 mg 250mg
30240712 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30240668 Supplier Toronto Research Chemicals Supplier No. TRCG1257502.5G
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10138-52-0
Molecular Formula Cl3 Gd
Molecular Weight (g/mol) 263.61
Synonym Gadolinium Chloride, NSC 174322
IUPAC Name trichlorogadolinium
SMILES Cl[Gd](Cl)Cl

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/3ClH.Gd/h3*1H;/q;;;+3/p-3
Formula Weight 262.8307
Chemical Name or Material Gadolinium Trichloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.