missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium Trichloride, TRC

Product Code. 30240998
Change view
Click to view available options
Quantity:
50 mg
250 mg
2.5 g
Unit Size:
2.5g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30240998 250 mg 250mg
30240668 2.5 g 2.5g
30240712 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30240998 Supplier Toronto Research Chemicals Supplier No. TRCG125750250MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10138-52-0
Molecular Formula Cl3 Gd
Molecular Weight (g/mol) 263.61
Synonym Gadolinium Chloride, NSC 174322
IUPAC Name trichlorogadolinium
SMILES Cl[Gd](Cl)Cl

Especificaciones

Quantity 250 mg
InChI Formula InChI=1S/3ClH.Gd/h3*1H;/q;;;+3/p-3
Formula Weight 262.8307
Chemical Name or Material Gadolinium Trichloride
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.