Learn More
Galactosylceramidase/GALC Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-33411-100ul
 View more versions of this product
Product Code. 30231152
  • 530.00€ / 100µL
Description
Galactosylceramidase/GALC Monoclonal antibody specifically detects Galactosylceramidase/GALC in Human samples. It is validated for ELISA,Immunocytochemistry/Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Galactosylceramidase/GALC
Monoclonal
ELISA Recommended starting concentration is 1 μg/mL, Immunocytochemistry/Immunofluorescence 1:50 - 1:200
EC 3.2.1.46, galactocerebrosidase, Galactocerebroside beta-galactosidase, Galactosylceramidase, galactosylceramidase (Krabbe disease), Galactosylceramide beta-galactosidase, galactosylceraminidase, GALCERase
Recombinant fusion protein containing a sequence corresponding to amino acids 40-300 of human Galactosylceramidase/GALC (NP_000144.2).,, Sequence:, GGAYVLDDSDGLGREFDGIGAVSGGGATSRLLVNYPEPYRSQILDYLFKPNFGASLHILKVEIGGDGQTTDGTEPSHMHYALDENYFRGYEWWLMKEAKKRNPNITLIGLPWSFPGWLGKGFDWPYVNLQLTAYYVVTWIVGAKRYHDLDIDYIGIWNERSYNANYIKILRKMLNYQGLQRVKIIASDNLWESISASMLLDAELFKVVDVIGAHYPGTHSAKDAKLTGKKLWSSEDFSTLNSDMGAGCWGRILNQNYINGY
100 μL
Cancer, Cardiovascular Biology, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction
2581
Store at -20°C. Avoid freeze-thaw cycles.
IgG
ELISA, Immunocytochemistry/Immunofluorescence
Unconjugated
PBS (pH 7.3), 50% glycerol, 0.05% BSA