Learn More
Biochrom Galapagos EZ Read PC Software
Product Code. 16835450 Shop All Harvard Bioscience Products
Change view
Click to view available options
For Use With (Equipment):
Microplate Reader
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
16835450 Microplate Reader Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16835450 Supplier Biochrom Supplier No. 80400241
  • ONLINE EXCLUSIVE PRICE
    1606.84€
    1550.50€ / Pack of 1
    Save 56.34€
    3% Off
Estimated Shipment: 15-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Galapagos EZ Read PC Software is intuitive, powerful and flexible software for all EZ Read Microplate Readers.
You don't need to be an expert to use EZ Read PC software: our intuitive approach makes it easy to add analysis to your measurement.
Designed for Elisa Assays:
  • Comprehensive formulae for built-in analysis of ELISA assays including curve-fitting options for 4 and 5 parameter fits-essential for many ELISA assays but difficult to do in Excel.
  • Easy to configure your plate layout depending on the number of samples: add multiple assays to a plate for analysing just a few samples or add multiple plates for analysing.
Designed for Research Labs:
  • Extensive offering of data analysis options for a range of absorbance assays.
  • Downloadable assay library for quick method setup, just click and drag your samples and go.
Uncomplicated Setup for Measurements and Analysis:
  • EZ Read PC software provides a clean and uncomplicated workspace to set up your analysis.
  • Intuitive step by step approach for creating an integrated measurement and analysis method.
  • Plate layout uses a simple 'click and drag' method for setting standard, control, sample and reference wells.
  • Plate definition editor has preinstalled plates with the capability to modify for specific plates for best measurement performance.
Specifications
Product Type Software for Microplate Reader
For Use With (Equipment) Microplate Reader