missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galectin-9 Antibody (1015214), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB20453MFV610
446.40 EUR valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
Galectin-9 Monoclonal antibody specifically detects Galectin-9 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF
Specifications
Galectin-9 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
0.1 mL | |
Myeloid derived Suppressor Cell, Signal Transduction | |
3965 | |
Store at 4C in the dark. |
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
1015214 | |
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
ecalectin, Gal-9, galectin 9, galectin-9, HUAT, lectin, galactoside-binding, soluble, 9, LGALS9A, MGC117375, MGC125973, MGC125974, Tumor antigen HOM-HD-21, urate transporter/channel protein | |
Human embryonic kidney cell, HEK293 derived human Galectin-9, Met1-Thr323, Accession # NP_002299 | |
RUO | |
Primary | |
Human | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction