missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Galectin 9 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifikationer
| Antigen | Galectin 9 |
|---|---|
| Concentration | 0.17 mg/mL |
| Applications | Immunofluorescence, Immunocytochemistry, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Produktkod | Brand | Quantity | Pris | Kvantitet och tillgänglighet | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkod | Brand | Quantity | Pris | Kvantitet och tillgänglighet | |||||
|
16837954
|
Proteintech
17938-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16827954
|
Proteintech
17938-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivning
The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. The protein encoded by this gene is an S-type lectin. It is overexpressed in Hodgkin′s disease tissue and might participate in the interaction between the H&RS cells with their surrounding cells and might thus play a role in the pathogenesis of this disease and/or its associated immunodeficiency. Multiple alternatively spliced transcript variants have been found for this gene.Specifikationer
| Galectin 9 | |
| Immunofluorescence, Immunocytochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| O00182 | |
| 3965 | |
| Galectin 9 Fusion Protein Ag12209 | |
| Primary | |
| -20°C | |
| LGALS9 |
| 0.17 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Ecalectin, Gal 9, Galectin 9, HUAT, LGALS9, LGALS9A, Tumor antigen HOM HD 21 | |
| LGALS9 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel