missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GALK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15164-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a highly efficient N-acetylgalactosamine kinase, which has galactokinase activity when galactose is present at high concentrations. Two alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
| GALK2 | |
| Polyclonal | |
| Unconjugated | |
| GALK2 | |
| galactokinase 2, GALK2, GalNAc kinase, GK2, N acetylgalactosamine kinase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2585, 296117, 69976 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q01415, Q5XIG6, Q68FH4 | |
| Galk2 | |
| GALK2 Fusion Protein Ag7278 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction