missing translation for 'onlineSavingsMsg'
Learn More

Gallium(III) iodide, ultra dry, 99.999% (metals basis), Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar 047148.03

Quantity 1g

 View more versions of this product

Product Code. 15457415

  • ONLINE EXCLUSIVE PRICE
    92.10€
    90.30€ / 1g
    Save 1.80€
    1% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

13450-91-4
450.436
DWRNSCDYNYYYHT-UHFFFAOYSA-K
83478
[Ga](I)(I)I
GaI3
MFCD00016107
gallium iii iodide, gallium iodide, gallium triiodide, acmc-20akty, gallium iodide,, gallium iii iodide, anhydrous, gallium iii iodide, ultra dry, gallium iii iodide trace metals basis, gallium iii iodide, anhydrous, powder trace metals basis
triiodogallane

Specifications

13450-91-4
GaI3
UN3260
Decomposes in water.
[Ga](I)(I)I
450.436
450.43
Ampule
4.15 g/mL
Gallium(III) iodide, Ultra dry
1g
MFCD00016107
gallium iii iodide, gallium iodide, gallium triiodide, acmc-20akty, gallium iodide,, gallium iii iodide, anhydrous, gallium iii iodide, ultra dry, gallium iii iodide trace metals basis, gallium iii iodide, anhydrous, powder trace metals basis
DWRNSCDYNYYYHT-UHFFFAOYSA-K
triiodogallane
83478
99.999%
Light and moisture sensitive
(metals basis)
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.