missing translation for 'onlineSavingsMsg'
Learn More

Gallium(III) iodide, ultra dry, 99.999% (metals basis), Thermo Scientific™

90.30€ - 606.98€

Chemical Identifiers

CAS 13450-91-4
Molecular Formula GaI3
Molecular Weight (g/mol) 450.436
MDL Number MFCD00016107
InChI Key DWRNSCDYNYYYHT-UHFFFAOYSA-K
Synonym gallium iii iodide, gallium iodide, gallium triiodide, acmc-20akty, gallium iodide,, gallium iii iodide, anhydrous, gallium iii iodide, ultra dry, gallium iii iodide trace metals basis, gallium iii iodide, anhydrous, powder trace metals basis
PubChem CID 83478
IUPAC Name triiodogallane
SMILES [Ga](I)(I)I
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15457415
SDS View Documents
Thermo Scientific Alfa Aesar
047148.03
1g
ONLINE EXCLUSIVE PRICE
92.10€
90.30€ / 1g
Save 1.80€
1% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15467415
SDS View Documents
Thermo Scientific Alfa Aesar
047148.06
5g
ONLINE EXCLUSIVE PRICE
183.00€
179.01€ / 5g
Save 3.99€
2% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15477415
SDS View Documents
Thermo Scientific Alfa Aesar
047148.14
25g
ONLINE EXCLUSIVE PRICE
619.00€
606.98€ / 25g
Save 12.02€
1% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Ampouled under argon
Specifications

Chemical Identifiers

13450-91-4
450.436
DWRNSCDYNYYYHT-UHFFFAOYSA-K
83478
[Ga](I)(I)I
GaI3
MFCD00016107
gallium iii iodide, gallium iodide, gallium triiodide, acmc-20akty, gallium iodide,, gallium iii iodide, anhydrous, gallium iii iodide, ultra dry, gallium iii iodide trace metals basis, gallium iii iodide, anhydrous, powder trace metals basis
triiodogallane

Specifications

13450-91-4
MFCD00016107
gallium iii iodide, gallium iodide, gallium triiodide, acmc-20akty, gallium iodide,, gallium iii iodide, anhydrous, gallium iii iodide, ultra dry, gallium iii iodide trace metals basis, gallium iii iodide, anhydrous, powder trace metals basis
DWRNSCDYNYYYHT-UHFFFAOYSA-K
triiodogallane
83478
99.999%
Light and moisture sensitive
(metals basis)
GaI3
UN3260
Decomposes in water.
[Ga](I)(I)I
450.436
450.43
Ampule
4.15 g/mL
Gallium(III) iodide, Ultra dry
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H314-H318-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.

P260-P264b-P271-P272-P280-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c

H314-H317-H334-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

missing translation for 'einecsNumber' : 236-611-6

missing translation for 'tsca' : Yes

missing translation for 'storageNote' :

  • Ampouled under argon

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.