missing translation for 'onlineSavingsMsg'
Learn More

(+)-Gallocatechin, TRC

Product Code. 17901735
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
1g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17901735 250 mg 250mg
17955025 50 mg 50mg
17944315 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17901735 Supplier Toronto Research Chemicals Supplier No. TRCG188995250MG
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 970-73-0
Molecular Formula C15H14O7
Molecular Weight (g/mol) 306.27
Synonym 2H-1-Benzopyran-3,5,7-triol, 3,4-dihydro-2-(3,4,5-trihydroxyphenyl)-, (2R-trans)-, Gallocatechol (8CI), Gallocatechol, d- (6CI), (2R,3S)-3,4-Dihydro-2-(3,4,5-trihydroxyphenyl)-2H-1-benzopyran-3,5,7-triol, (+)-Gallocatechin, (+)-Gallocatechol, (2R,3S)-(+)-Gallocatechin, Gallocatechin, NSC 674038, Sunphenon EGC-OP, d-Gallocatechin, Gallocatechol, (+)-gallocatechol, D-Gallocatechin;
IUPAC Name (2R,3S)-2-(3,4,5-trihydroxyphenyl)-3,4-dihydro-2H-chromene-3,5,7-triol
SMILES O[C@H]1Cc2c(O)cc(O)cc2O[C@@H]1c3cc(O)c(O)c(O)c3

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C15H14O7/c16-7-3-9(17)8-5-12(20)15(22-13(8)4-7)6-1-10(18)14(21)11(19)2-6/h1-4,12,15-21H,5H2/t12-,15+/m0/s1
Formula Weight 306.074
Percent Purity >95%
Chemical Name or Material (+)-Gallocatechin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.