missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GALNT4 Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5149364
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
This gene encodes a member of the UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase (GalNAc-T) family of enzymes. GalNAc-Ts initiate mucin-type O-linked glycosylation in the Golgi apparatus by catalyzing the transfer of GalNAc to serine and threonine residues on target proteins. They are characterized by an N-terminal transmembrane domain, a stem region, a lumenal catalytic domain containing a GT1 motif and Gal/GalNAc transferase motif, and a C-terminal ricin/lectin-like domain. GalNAc-Ts have different, but overlapping, substrate specificities and patterns of expression. In vitro, the encoded protein can complement other GalNAc-Ts in the complete O-glycosylation of the mucin-1 tandem repeat and can O-glycosylate the P-selectin glycoprotein ligand-1 molecule. The coding region of this gene is contained within a single exon.
Specifications
| GALNT4 | |
| Polyclonal | |
| Unconjugated | |
| GALNT4 | |
| AV011803; GALNACT4; GalNAc-T4; Galnt4; Polypeptide GalNAc transferase 4; polypeptide GalNAc transferase-T4; polypeptide N-acetylgalactosaminyltransferase 4; pp-GaNTase 4; ppGaNTase-T4; protein-UDP acetylgalactosaminyltransferase 4; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 4; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 4; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 4 (GalNAc-T4) | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 8693 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q8N4A0 | |
| GALNT4 | |
| Mouse myeloma cell line NS0-derived recombinant human Polypeptide GalNac Transferase 4/GALNT4, Ala36-Lys578. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction