missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GALNT5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
292.00€ - 572.00€
Specifications
| Antigen | GALNT5 |
|---|---|
| Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18495621
|
Novus Biologicals
NBP1-86484-25ul |
25 μL |
292.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18225466
|
Novus Biologicals
NBP1-86484 |
0.1 mL |
572.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GALNT5 Polyclonal specifically detects GALNT5 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| GALNT5 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| EC 2.4.1.41, GALNAC-T5, MGC165041, Polypeptide GalNAc transferase 5, polypeptide N-acetylgalactosaminyltransferase 5, pp-GaNTase 5, Protein-UDP acetylgalactosaminyltransferase 5, UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 5, UDP-N-acetyl-alpha-D-galactosamine:polypeptideN-acetylgalactosaminyltransferase 5 (GalNAc-T5) | |
| GALNT5 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 11227 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:AERDLNVTISLSTDRPKQRSQAVANERAHPASTAVPKSGEAMALNKTKTQSKEVNANKHKANTSLPFPKFTVNSNRLRKQSINETPLGSLSKDDGARGAHGKKLNFSESHLVIITKEEEQKADPKEVSNSKTKTIFPKVLGKS | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title