missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GALNTL2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | GALNTL2 |
|---|---|
| Concentration | 0.13 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16856664
|
Proteintech
16900-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16846664
|
Proteintech
16900-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The function of this protein remains unknown.Specifications
| GALNTL2 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q8N3T1 | |
| 117248 | |
| GALNTL2 Fusion Protein Ag10434 | |
| Primary | |
| -20°C | |
| GALNT15 |
| 0.13 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| GalNAc T like protein 2, GALNT15, GALNT7, GALNTL2, PIH5, pp GaNTase like protein 2 | |
| GALNT15 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title