Learn More
gamma C Crystallin Antibody (OTI1E3), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44646
This item is not returnable.
View return policy
Description
gamma C Crystallin Monoclonal antibody specifically detects gamma C Crystallin in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| gamma C Crystallin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized powder ( 1X PBS, pH 7.3,) | |
| Mouse | |
| >95% | |
| RUO | |
| Primary | |
| For reconstitution, we recommend adding 100uL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiment, we strongly recommend performing another round of desalting process. | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| OTI1E3 | |
| Western Blot 1:2000 | |
| CCL, CRYG3gamma-C-crystallin, crystallin, gamma C, crystallin, gamma-3, Gamma-C-crystallin, Gamma-crystallin 2-1, Gamma-crystallin 3, gamma-crystallin C | |
| Full length human recombinant protein of human CRYGC (NP_066269) produced in HEK293T cell. | |
| 100 μg | |
| Vision | |
| 1420 | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2a |