Learn More
Gamma-Terpinene, TRC
Product Code. 30428311
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30428311 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30428311 Supplier Toronto Research Chemicals Supplier No. TRCT11750325G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material gamma-Terpinene
CAS 99-85-4
Purity Grade Notes GC
Molecular Formula C10 H16
Quantity 25 g
Synonym 1,4-Cyclohexadiene','1-methyl-4-(1-methylethyl)-, p-Mentha-1,4-diene (8CI), 1-Methyl-4-(1-methylethyl)-1,4-cyclohexadiene, 1-Isopropyl-4-methyl-1,4-cyclohexadiene, 1-Methyl-4-isopropyl-1,4-cyclohexadiene, 4-Isopropyl-1-methyl-1,4-cyclohexadiene, Crithmene, Moslene, NSC 21448, gamma-Terpinene, γ-Terpinen, γ-Terpinene
InChI Formula InChI=1S/C10H16/c1-8(2)10-6-4-9(3)5-7-10/h4,7-8H,5-6H2,1-3H3
SMILES CC(C)C1=CCC(=CC1)C
IUPAC Name 1-methyl-4-propan-2-ylcyclohexa-1,4-diene
Molecular Weight (g/mol) 136.23
Formula Weight 136.1252
Percent Purity >95