missing translation for 'onlineSavingsMsg'
Learn More

Gamma-Tocopherol-d4 (major), TRC

Product Code. 30457208
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457208 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30457208 Supplier Toronto Research Chemicals Supplier No. TRCT5261421MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material gamma-Tocopherol-d4
CAS 1329652-13-2
Purity Grade Notes HPLC
Molecular Formula C28H44D4O2
Quantity 1 mg
Synonym (2R)-3,4-Dihydro-2,8-dimethyl-7-(methyl-d4)-2-[(4R,8R)-4,8,12-trimethyltridecyl]-2H-1-benzopyran-6-ol, (+)- 2,8-Dimethyl-(7-methyl-d4)-2-(4,8,12-trimethyltridecyl) -6-chromanol, (+)-γ-Tocopherol-d4, (2R,4'R,8'R)-γ-Tocopherol-d4, (R,R,R)-γ -Tocopherol-d4, D-γ-Tocopherol-d4, all-(R)-γ-Tocopherol-d4, d-γ-Tocopherol-d4
InChI Formula InChI=1S/C28H48O2/c1-20(2)11-8-12-21(3)13-9-14-22(4)15-10-17-28(7)18-16-25-19-26(29)23(5)24(6)27(25)30-28/h19-22,29H,8-18H2,1-7H3/t21-,22-,28-/m1/s1/i5D3,19D
SMILES OC1=C([2H])C(CC[C@](CCC[C@H](C)CCC[C@H](C)CCCC(C)C)(C)O2)=C2C(C)=C1C([2H])([2H])[2H]
Molecular Weight (g/mol) 420.7
Formula Weight 420.3905
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.