missing translation for 'onlineSavingsMsg'
Learn More
Learn More
gamma Tubulin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
227.00€ - 326.00€
Specifications
| Antigen | gamma Tubulin |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18619625
|
Novus Biologicals
NBP2-99067-100ul |
100 μg |
326.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18602086
|
Novus Biologicals
NBP2-99067-50ul |
50 μg |
227.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
gamma Tubulin Polyclonal antibody specifically detects gamma Tubulin in Human samples. It is validated for Western BlotSpecifications
| gamma Tubulin | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cellular Markers, Centrosome Markers | |
| 0.2 μm filtered solution in PBS | |
| 7283 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| gamma-1-tubulin, Gamma-tubulin complex component 1, GCP-1, TUBG, TUBGCP1, tubulin gamma-1 chain, tubulin, gamma 1, tubulin, gamma polypeptide | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human gamma Tubulin. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title