Learn More
BW Technologies™ Gas Detectors
Product Code. 11960917
Change view
Click to view available options
Components Measured:
Oxygen, Carbon Monoxide, Hydrogen Sulfide, Sulfur Dioxide, VOC, Carbon Dioxide Gas and Fuel
Oxygen, LEL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Components Measured Unit Size
11960917 Oxygen, Carbon Monoxide, Hydrogen Sulfide, Sulfur Dioxide, VOC, Carbon Dioxide Gas and Fuel Each
11968995 Oxygen, LEL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 11960917 Supplier BW Technologies™ Supplier No. M5IRXWBYRDDYE00
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Specifications
Alarm Types Audible
Battery Life 2 Years
Battery Type 2 AA
Certifications/Compliance Ex II 1 G T4 Eex ia IIC T4 except CO2 detector
Depth (Metric) 38 mm
Components Measured Oxygen, Carbon Monoxide, Hydrogen Sulfide, Sulfur Dioxide, VOC, Carbon Dioxide Gas and Fuel
Type Gas Detector
Height (Metric) 145 mm
IP Rating IP67
Length (Metric) 74 mm
Weight (Metric) 370g