Learn More
3M™ Gas and Vapor Cartridge, 6000 Series
Product Code. 11783343
Change view
Click to view available options
:
Model: 6051; Filter type: A1
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11783343 Model: 6051; Filter type: A1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11783343 Supplier 3M™ Supplier No. 7000034751
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Combines excellent protection with superb comfort and balance. The 3M Gas and Vapor Cartridge is trapezoidal shaped and works with half- and full-facemasks that have bayonet connections systems.
Each filter has a colored band using the EN14387 classification system, to denote the nature of the protection it provides.
  • Trapezoidal-shaped gas and vapor filters combine excellent protection with superb comfort and balance
  • Lightweight, low breathing resistance, and well balanced when fitted to appropriate mask
  • Provides excellent field of vision as a result of the unique trapezoidal shape.
  • Filters can be used on 3M™ Facemasks fitted with a bayonet connections system
TRUSTED_SUSTAINABILITY
Specifications
Cartridge Type A1
Certifications/Compliance EN14387:2004 + A1:2008
Connection Type Bayonet
Description Organic Vapors bp>65°C
Product Type Gas and Vapor Cartridge
For Use With (Application) Anywhere conventional paints are used (subject to usage conditions), Vehicle manufacture, Aircraft manufacture & refurbishment, Ink & dye manufacturer and use, Adhesive manufacturer and use, Paint and varnish manufacturer, Resin manufacturer and use
For Use With (Equipment) Half and Full Facemasks
Quantity 8/Pk.