Learn More
3M™ Gas and Vapor Cartridge, 6000 Series
Description
Each filter has a colored band using the EN14387 classification system, to denote the nature of the protection it provides.
- Trapezoidal-shaped gas and vapor filters combine excellent protection with superb comfort and balance
- Lightweight, low breathing resistance, and well balanced when fitted to appropriate mask
- Provides excellent field of vision as a result of the unique trapezoidal shape.
- Filters can be used on 3M™ Facemasks fitted with a bayonet connections system
Specifications
Specifications
| Cartridge Type | A1 |
| Certifications/Compliance | EN14387:2004 + A1:2008 |
| Connection Type | Bayonet |
| Description | Organic Vapors bp>65°C |
| Product Type | Gas and Vapor Cartridge |
| For Use With (Application) | Anywhere conventional paints are used (subject to usage conditions), Vehicle manufacture, Aircraft manufacture & refurbishment, Ink & dye manufacturer and use, Adhesive manufacturer and use, Paint and varnish manufacturer, Resin manufacturer and use |
| For Use With (Equipment) | Half and Full Facemasks |
| Quantity | 8/Pk. |