Learn More
3M™ Gas and Vapour Cartridge 6054
Product Code. 15729667
Change view
Click to view available options
:
Quantity: 64/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15729667 Quantity: 64/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15729667 Supplier 3M™ Supplier No. 7000030095
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
3M™ Gas and Vapour Cartridge 6054 protect against ammonia vapours.
  • K1 Filters
  • Suitable for 3M™ 6000 and 3M™ 7000 Series Half and Full Face Masks
TRUSTED_SUSTAINABILITY
Specifications
Cartridge Type K1
Certifications/Compliance EN14387:2004 + A1:2008
Description 6054 Filter
Filter Type Gas and Vapour Cartridge
Quantity 64/Cs.