Learn More
3M™ Gas and Vapour Cartridge 6054
Description
- K1 Filters
- Suitable for 3M™ 6000 and 3M™ 7000 Series Half and Full Face Masks
Specifications
Specifications
| Cartridge Type | K1 |
| Certifications/Compliance | EN14387:2004 + A1:2008 |
| Description | 6054 Filter |
| Filter Type | Gas and Vapour Cartridge |
| Quantity | 64/Cs. |