missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAS7 Antibody (4H8), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-30203V
This item is not returnable.
View return policy
Description
GAS7 Monoclonal antibody specifically detects GAS7 in Human samples. It is validated for Western Blot, ELISA
Specifications
| GAS7 | |
| Monoclonal | |
| DyLight 405 | |
| NP_958836 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 4H8 | |
| 50mM Sodium Borate | |
| GAS-7, growth arrest-specific 7, KIAA0394growth arrest-specific protein 7, MGC1348, MLL/GAS7, MLL/GAS7 fusion protein | |
| Recombinant human GAS7 (1-416aa) purified from E. coli | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 8522 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction