missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GATA-3 Antibody (GATA3/7686R), Novus Biologicals™
Rabbit Monoclonal Antibody
268.00€ - 496.00€
Specifications
| Antigen | GATA-3 |
|---|---|
| Clone | GATA3/7686R |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin : 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18603665
|
Novus Biologicals
NBP3-23826-20ug |
20 μg |
268.00€
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18619944
|
Novus Biologicals
NBP3-23826-100ug |
100 μg |
496.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GATA-3 Monoclonal antibody specifically detects GATA-3 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| GATA-3 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Innate Immunity | |
| 10 mM PBS with 0.05% BSA | |
| 2625 | |
| IgG κ | |
| Protein A or G purified |
| GATA3/7686R | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 131320, GATA binding protein 3, GATA-binding factor 3, GATA-binding protein 3, HDRMGC5199, MGC2346, MGC5445, trans-acting T-cell-specific transcription factor GATA-3 | |
| Recombinant fragment (around aa150-250) of human GATA-3 protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title