Learn More
Invitrogen™ Gatralimab Recombinant Monoclonal Antibody

Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA559237
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Gatralimab is a biosimilar that targets CD52. Human CD52, also known as Cambridge pathology antigen 1 (CAMPATH-1), is a glycoprotein found in the male reproductive tract and immune cells. It is abundantly expressed on the surface of nearly all human peripheral blood lymphocytes. The CD52 antigen is Gpi-anchored membrane receptor that consists of a small 12 amino acid peptide attached to a large N-linked carbohydrate group. CD52 is a potent target for inducing cytolysis of mature lymphocytes. Alemtuzumab, a humanized monoclonal antibody targeting CD52, is used as an immunomodulatory drug for the treatment of chronic lymphocytic leukemia, T cell lymphomas, multiple sclerosis, and other diseases.
Specifications
| Gatralimab Humanized | |
| Recombinant Monoclonal | |
| Unconjugated | |
| GZ-402668 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Human | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.