missing translation for 'onlineSavingsMsg'
Learn More

GB 31660.6-2019 α2-Agonists Mixture 169 100 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30328560
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30328560 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30328560 Supplier Dr. Ehrenstorfer Supplier No. DREA50000169ME

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Apraclonidine Hydrochloride∣Brimonidine∣Clonidine Hydrochloride∣Tizanidine hydrochloride∣Xylazine
CAS 73218-79-8, 59803-98-4, 4205-91-8, 64461-82-1, 7361-61-7
Analyte or Component Names Apraclonidine Hydrochloride∣Brimonidine∣Clonidine Hydrochloride∣Tizanidine hydrochloride∣Xylazine
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Methanol
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Apraclonidine Hydrochloride 100 μg/mL∣Brimonidine 100 μg/mL∣Clonidine Hydrochloride 100 μg/mL∣Tizanidine hydrochloride 100 μg/mL∣Xylazine 100 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.