missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GBP1 Antibody (GBP1/7617) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-26937JF525
This item is not returnable.
View return policy
Description
GBP1 Monoclonal antibody specifically detects GBP1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| GBP1 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| GBP-1, GTP-binding protein 1, Guanine nucleotide-binding protein 1, guanylate binding protein 1, interferon-inducible, guanylate binding protein 1, interferon-inducible, 67kDa, huGBP-1, interferon-induced guanylate-binding protein 1 | |
| Recombinant fragment (around aa400-592) of human GBP1 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Immunology, Innate Immunity, Signal Transduction | |
| 2633 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| GBP1/7617 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction