missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GBX2 Antibody (GBX2/7235), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24331MFV610
This item is not returnable.
View return policy
Description
GBX2 Monoclonal antibody specifically detects GBX2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
GBX2 | |
Monoclonal | |
mFluor Violet 610 SE | |
Gastrulation and brain-specific homeobox protein 2, gastrulation brain homeo box 2, gastrulation brain homeobox 2, homeobox protein GBX-2 | |
Recombinant fragment (around aa150-350) of human GBX2 protein (exact sequence is proprietary) | |
0.1 mL | |
Neuroscience | |
2637 | |
Store at 4°C in the dark. | |
IgG2 κ |
Immunohistochemistry (Paraffin) | |
GBX2/7235 | |
50 mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction