Learn More
SGE Analytical Science™ GC Autosampler Syringe
Description
- AutoSystem™ and Clarus™ Removable Needle
- 0.5μL syringe
- FOR PE
- 0.63mm O.D.
Specifications
Specifications
| Needle Gauge | 23 G |
| Diameter (Metric) Outer Needle | 0.63 mm |
| Product Type | Autosampler Syringe |
| Volume (Metric) | 0.5 μL |
| Needle Point Style | Cone |