missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCM1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-48520-25ul
This item is not returnable.
View return policy
Description
GCM1 Polyclonal antibody specifically detects GCM1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| GCM1 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL | |
| GCM motif protein 1, GCMA, glial cells missing (Drosophila) homolog a, Glial cells missing homolog 1, glial cells missing homolog 1 (Drosophila), hGCMachorion-specific transcription factor GCMa | |
| This antibody was developed against a recombinant protein corresponding to amino acids: LGRNHLADNCYSNYPFPLTSWPCSFSPSQNSSEPFYQQLPLEPPAAKTGCPPLWPNPAGNLYEEKVHVDFNSYVQSPAYHSPQEDPFLFTYASHPHQQYSLPSKSSKWDFEEEMTYLGLDHCNNDMLLNLCP | |
| 25 μL | |
| Primary | |
| Human | |
| Purified |
| Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 8521 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction