missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GCSH Recombinant Rabbit Monoclonal Antibody (5U2H0)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA558156

 View more versions of this product

Product Code. 30283725

  • 457.00€ / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: SVRKFTEKHE WVTTENGIGT VGISNFAQEA LGDVVYCSLP EVGTKLNKQD EFGALESVKA ASELYSPLSG EVTEINEALA ENPGLVNKSC YEDGWLIKMT LSNPSELDEL MSEEAYEKYI KSIEE.

The glycine cleavage system catalyzes the degradation of glycine. The H protein (GCSH) shuttles the methylamine group of glycine from the P protein (GLDC) to the T protein (GCST).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GCSH
Recombinant Monoclonal
1 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
P23434
GCSH
Recombinant fusion protein containing a sequence corresponding to amino acids 49-173 of human GCSH (NP_004474.2).
100 μL
Primary
Human
Antibody
IgG
ELISA, Immunocytochemistry
5U2H0
Unconjugated
GCSH
1100001L02Rik; 5730591C18Rik; GCE; Gcsh; Glycine cleavage system H protein, mitochondrial; glycine cleavage system protein H; glycine cleavage system protein H (aminomethyl carrier); lipoic acid-containing protein; mitochondrial glycine cleavage system H-protein; NKH
Rabbit
Protein A
RUO
2653
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.