missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GDF15 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27455-1-AP-150UL
This item is not returnable.
View return policy
Description
Bone morphogenetic proteins (e. g., BMP5, MIM 112265) are members of the transforming growth factor-beta (see TGFB1, MIM 190180) superfamily and regulate tissue differentiation and maintece. They are synthesized as precursor molecules that are processed at a dibasic cleavage site to release C-terminal domains containing a characteristic motif of 7 conserved cysteines in the mature protein.Specifications
| GDF15 | |
| Polyclonal | |
| Unconjugated | |
| GDF15 | |
| GDF 15, GDF15, MIC 1, MIC1, NAG 1, NRG 1, NSAID activated gene 1 protein, NSAID regulated gene 1 protein, PDF, PLAB, Placental TGF beta, PTGFB | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23886, 9518 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q99988, Q9Z0J7 | |
| GDF15 | |
| GDF15 Fusion Protein Ag26760 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction