missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GDF3 Monoclonal Antibody (A7), Invitrogen™

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA543806
This item is not returnable.
View return policy
Description
GDF3 is a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. The function of this protein is unknown, but expression studies suggest it may be involved in regulation of the adult lymphatic and erythroid systems and embryonic development.
Specifications
| GDF3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q9NR23 | |
| GDF3 | |
| Ala251-Gly364 (Accession Q9NR23), with N-terminal His Tag. | |
| 100 μL | |
| Primary | |
| Human, Rat, Pig | |
| Antibody | |
| IgG2b κ |
| Western Blot, Immunocytochemistry | |
| A7 | |
| Unconjugated | |
| GDF3 | |
| C78318; ecat9; Gdf3; Gdf-3; growth differentiation factor 3; growth/differentiation factor 3; KFS3; MCOP7; MCOPCB6; RGD1564178; UNQ222/PRO248; VG-1-related protein 2; Vgr2; Vgr-2 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 100510963, 500311, 9573 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction