missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GDI2 Antibody (0B10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-42973
This item is not returnable.
View return policy
Description
GDI2 Monoclonal antibody specifically detects GDI2 in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| GDI2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| 0B10 | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry 1:20 - 1:200, Immunocytochemistry/ Immunofluorescence 1:10 - 1:100 | |
| beta, FLJ16452, GDP dissociation inhibitor 2, Guanosine diphosphate dissociation inhibitor 2, RABGDIBFLJ37352 | |
| GDP dissociation inhibitor 2 - (Uniprot# P50395) | |
| 100 μg | |
| Apoptosis, Signal Transduction | |
| 2665 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction