missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Gedunin

Product Code. 18762734
Change view
Click to view available options
Quantity:
2 mg
Unit Size:
2mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18762734 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18762734 Supplier Tocris Bioscience™ Supplier No. 3387/2

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Hsp90 inhibitor; exhibits anticancer and antimalarial activity

Gedunin is a naturally occurring Hsp90 inhibitor. Induces Hsp90-dependent client protein degradation and displays antiproliferative activity in vitro(IC50 values are 3.22, 8.84 and 16.8 μM in SKBr3, MCF-7 and CaCo-2 cancer cell lines respectively). Decreases ovarian cancer cell proliferation in vitroand may enhance cisplatin -induced reduction in proliferation (CAT.No 3387). Also exhibits antimalarial activity against Plasmodium falciparum(IC50 values are 0.14 and 3.1 μM in parasite development and [3H]-hypoxanthine uptake assays respectively).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (1S,3aS,4aR,4bS,5R,6aR,10aR,10bR,12aS)-5-(Acetyloxy)-1-(3-furanyl)-1,5,6,6a,7,10a,10b,11,12,12a,decahydro-4b,7,7,10a,12a,-pentamethyloxireno[c]phenanthro[1,2-d]pyran-3,8(3aH,4bH)-dione
CAS 2753-30-2
Quantity 2 mg
Target HSP90 Inhibitors
Molecular Formula C28H34O7
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.