missing translation for 'onlineSavingsMsg'
Learn More

GEFT Antibody (7A1), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-42974

Product Code. 30279368

  • 408.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

GEFT Monoclonal antibody specifically detects GEFT in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GEFT
Monoclonal
Unconjugated
PBS, pH 7.3, 50% glycerol
Mouse
Protein A or G purified
RUO
115557
Store at -20°C. Avoid freeze-thaw cycles.
IgG1
ELISA, Immunohistochemistry, Western Blot
7A1
Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry 1:20 - 1:200
GEFTRAC/CDC42 exchange factor, Guanine nucleotide exchange factor GEFT, p63RhoGEFrho guanine nucleotide exchange factor 25, Rac/Cdc42/Rho exchange factor GEFT, Rho guanine nucleotide exchange factor (GEF) 25, RhoA/RAC/CDC42 exchange factor, RhoA/Rac/Cdc42 guanine nucleotide exchange factor GEFT
RhoA/RAC/CDC42 exchange factor - (Uniprot# Q86VW2)
100 μg
Primary
Human, Mouse, Rat
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.