Learn More
Fisherbrand™ Gel Casting Tray
Product Code. 11563352 Shop All Fisherbrand Electrochemistry Products
Change view
Click to view available options
Dimensions (L x W):
140 x 230 mm
144 x 102 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
11563352 144 x 102 mm Each
11583382 140 x 230 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11563352
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Accessory for use with Fisherbrand Wide Format Horizontal Gel Units.
  • Removable
  • Designed with gates
Specifications
Description Removable
For Use With (Equipment) Wide Format Mini-Plus Horizontal Gel Unit
Type Gel Casting Tray
Includes Gates
Dimensions (L x W) 144 x 102 mm