missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gelatin (Bovine), TRC
Specifications
Specifications
| CAS | 9000-70-8 |
| Format | Neat |
| Quantity | 25 g |
| Shipping Condition | Room Temperature |
| Synonym | Gelatins∣Gelatin∣Gelatins, acetylated, conjugates∣Gelatins, conjugates∣225 Bloom∣250 Bloom∣260B∣260B (gelatin)∣A 1956∣A 330∣A-U∣AP 100∣AP 150∣AP 150 (gelatin)∣AP 200∣AP 200 (gelatin)∣AP 250∣AP 250 (gelatin)∣AP 30∣AP 30 (gelatin)∣APB-H∣APH 100∣APH 150∣APH 200∣APH 250∣APH 250TS∣AU-P∣Bloom 0∣Bloom 180∣Bloom 220∣Bloom 225∣Bloom 240∣Bloom 250∣Bloom 280∣Bloom 300∣Bloom 302∣Bloom 330∣Bloom Strength 185∣CLV∣CLW∣CP∣CP (gelatin)∣Collagens, gelatins∣Conjugates acetylated gelatins∣Conjugates, gelatins∣Cryogel 220/440∣DAB 7∣DAB 7 (gelatin)∣E 200∣E 200 (gelatin)∣E 260∣E 290∣E 300∣E 300 (gelatin)∣EGDI 1∣Emagel∣F 275∣F 275 (gelatin)∣FCR 500∣FGH 2001∣FGL 200TS∣FGL 250∣FGL 250TS∣FGS 260∣FYB 150S∣FYB 200S∣FYB 250S∣FreAlagin AD∣FreAlagin M∣G 0282K∣G 0283K∣G 0284K∣G 0545P∣G 0637K∣G 0665K∣G 0666K∣G 0667K∣G 0668K∣G 0701K∣G 0723K∣G 0746K∣G 0749K∣G 0750∣G 0856K∣G 0866K∣G 0886K∣G 1171K∣G 1221K∣G 1642P∣G 1890∣G 3038E∣G 6650∣G 7041∣G 9382∣G 9391∣G Bifun∣GBL 100∣GBL 100 (gelatin)∣GBL 150∣GBL 200∣GBL 250∣GE 3V∣GLD 2∣GLD 3∣GPA-A 0∣GSN∣GX 45L404∣GXAS 7∣Gel Up J 3557∣Gel-Lac∣Gelafusal∣Gelatin 200∣Gelatin AU-P∣Gelatin Bloom 30∣Gelatin NNH∣Gelatin P 510∣Gelatin conjugates∣Gelatine∣Gelatine Bloom 250∣Gelfoam∣Gelita Bloom 302∣Gelita Collagel BS∣Gelita Sol E∣Gelita T 7838∣Gelita-Collagel∣Gelita-Sol P∣Gelita-Tec∣Gelodan G∣Gelofusin∣Gelofusine∣Geltec UG∣Glutins (gelatins)∣Grenetine∣Grindsted G∣HK 27∣IJ-SKW∣IK∣IK (gelatin)∣IK 195∣IK 2000∣IK 3000∣IXOS Gel FGL 200SP∣IXOS Gel FGL 200TS∣IXOS Gel HDL 50SP∣Inagel A 83∣Inagel AP 81P∣Inagel N 150∣JFG 230∣JS 110∣JS 190∣Jellice∣K 16096∣K 7598∣K 911216∣K Jelly∣KPF 16353∣KV 3000∣KV 3000 (gelatin)∣KV 3029∣LP 250∣LP 250 (gelatin)∣LS-W∣M 394∣M 396∣M 400∣M 400 (gelatin)∣MGP 9066∣MRK∣Medigelatin∣Medigelatin HMG-BP∣N 1236∣Neosoft GE 388∣Neosoft GE 82∣Nikkol CCP 4∣Nitta 750∣Nitta 810∣Nitta G 2426P∣Nitta MJ∣Nitta RR∣Nittait GF 600A∣Novotec 100∣Nyushiruba Karyu∣Oetker∣Ossein Gelatin HOS 1∣P 100∣P 100 (gelatin)∣P 104∣P 2115∣P 2225∣P 487∣PA 10∣PA 10 (gelatin)∣PA 100∣PA 100 (peptide)∣PB Leiner∣PM 46D∣PM 64∣PM 64 (gelatin)∣PS 100∣PS 100 (gelatin)∣PSK Gelatin∣PSP Gelatin∣PZ (gelatin)∣Pharmagel A∣Pharmagel AdB∣Pharmagel B∣Plantis RD∣Plantis RD Fine∣Plantis WT∣Polypro 5000∣Pulviclar S∣RBM 11BML∣RGB∣RM 100B∣Rousselot 160LB∣Rousselot 200FG30∣Rousselot 200PS∣Rousselot 3046∣Rousselot 53676∣Rousselot G.U.B.E.∣Rousselot SAS∣S 1286∣S 510∣S 510 (gelatin)∣S 511-5∣S 692∣S-A 354-1∣SB-GEL SLW∣SN∣SN (protein)∣SRC∣SRC (gelatin)∣Spongel∣Spongiofort∣Super Gelatin SSB∣Surgiflo∣Surgifoam∣TS 361PC∣VYSE∣ViniGel FL∣Vitrigel∣Z-KN 707∣beMatrix Gelatin LS-H∣beMatrix LS-H∣iXOS HDL 30DR |
| Chemical Name or Material | Gelatin |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?