Learn More
Gelatine, Extra Pure, SLR, Granular Powder, meets analytical specification of Ph.Eur., Fisher Chemical™
Shop All Fisher Chemical ProductsSpecifications
Specifications
| Color | Beige |
| pH | 5 to 7 |
| Packaging | Plastic powder jar |
| Quantity | 250 g |
| Physical Form | Powder |