Learn More
R&D Systems™ Geldanamycin
Product Code. 18646528 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18646528 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18646528 Supplier R&D Systems™ Supplier No. NB11081559
  • ONLINE EXCLUSIVE PRICE
    363.43€
    302.40€ / 1mg
    Save 61.03€
    16% Off
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
A Novus product, part of R&D Systems.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at -20°. Avoid freeze/thaw cycles.
Product Line Novus
Quantity 1 mg
For Use With (Application) Yellow Solid. Slightly Soluble in Methanol, Chloroform or DMSO (10 mg/mL); Insoluble aq. Soln..
Type Geldanamycin