Description
- One side is designed for gels of size: Full size (260 x 125mm), IEF: 10.4mL, All other: 25mL
- Other side is for smaller gels or PhastGel media: Half size (130 x 125mm), IEF: 5.2mL; All other: 13mL
Procedure:
- Select appropriate reswelling chamber of GelPool, depending on gel size
- Clean GelPool thoroughly with distilled water and let it dry. Do not use organic solvents
- Pipette selected gel rehydration solution along left edge of gel tray
Specifications
Specifications
| For Use With (Application) | Rehydration of dry gels of different sizes |
| Type | GelPool |
| Quantity | 1/Pk. |