missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gelsolin Mouse anti-Human, Clone: 3C4A2, Proteintech
Mouse Monoclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | Gelsolin |
|---|---|
| Clone | 3C4A2 |
| Concentration | 1 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16808015
|
Proteintech
66280-1-IG-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16898005
|
Proteintech
66280-1-IG-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene binds to the "plus" ends of actin monomers and filaments to prevent monomer exchange. The encoded calcium-regulated protein functions in both assembly and disassembly of actin filaments. Defects in this gene are a cause of familial amyloidosis Finnish type (FAF). Multiple transcript variants encoding several different isoforms have been found for this gene.Specifications
| Gelsolin | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Actin depolymerizing factor, ADF, AGEL, Brevin, Gelsolin, GSN | |
| GSN | |
| IgA | |
| Affinity Chromatography | |
| Antibody |
| 3C4A2 | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| P06396 | |
| 2934 | |
| Gelsolin Fusion Protein Ag17916 | |
| Primary | |
| -20°C | |
| GSN |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title