missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Gemfibrozil, 98%

Product Code. 15723457 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15723457 25 g 25g
15713457 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15723457 Supplier Thermo Scientific Chemicals Supplier No. 461790250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 25812-30-0 | C15H22O3 | 250.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25812-30-0
Molecular Formula C15H22O3
Molecular Weight (g/mol) 250.34
MDL Number MFCD00079335
InChI Key HEMJJKBWTPKOJG-UHFFFAOYSA-N
Synonym gemfibrozil, lopid, 5-2,5-dimethylphenoxy-2,2-dimethylpentanoic acid, jezil, decrelip, lipur, bolutol, apo-gemfibrozil, gemfibromax, cholespid
PubChem CID 3463
ChEBI CHEBI:5296
IUPAC Name 5-(2,5-dimethylphenoxy)-2,2-dimethylpentanoic acid
SMILES CC1=CC=C(C)C(OCCCC(C)(C)C(O)=O)=C1

Specifications

Melting Point 60.0°C to 61.0°C
Quantity 25 g
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 250.33
Percent Purity 98%
Chemical Name or Material Gemfibrozil

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.