Learn More
GEMIN8 Monoclonal Antibody (OTI4E11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805831S
Description
GEMIN8 Monoclonal antibody specifically detects GEMIN8 in Human samples. It is validated for Western BlotSpecifications
| GEMIN8 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O43809 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 11051 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI4E11 | |
| Unconjugated | |
| NUDT21 | |
| B130034M20Rik; B930082C09Rik; BC023488; Fam51a1; family with sequence similarity 51, member A1; family with sequence similarity 51, member A1 homolog; gem (nuclear organelle) associated protein 8; gem nuclear organelle associated protein 8; gem-associated protein 8; GEMIN8; Gemin-8; Protein FAM51A1 | |
| Human recombinant protein fragment corresponding to amino acids 2-227 of human NUDT21 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.