Learn More
- Home
- Products
- Humidity and Hygrometry
- Hydrometers
- Alla France™ General Purpose Density Hydrometer
Alla France™ General Purpose Density Hydrometer
Product Code. 11743193
Change view
Click to view available options
Specific Gravity Range:
0.140 to 0.150 g/mL
0.150 to 0.160 g/mL
0.160 to 0.170 g/mL
0.170 to 0.180 g/mL
0.180 to 0.190 g/mL
0.600 to 0.700 g/mL
0.700 to 0.800 g/mL
0.800 to 0.900 g/mL
0.850 to 0.900 g/mL
0.900 to 0.950 g/mL
0.900 to 1.000 g/mL
1.050 to 1.100 g/mL
1.200 to 1.250 g/mL
1.250 to 1.300 g/mL
1.300 to 1.350 g/mL
1.300 to 1.400 g/mL
1.350 to 1.400 g/mL
1.400 to 1.450 g/mL
1.400 to 1.500 g/mL
1.500 to 1.600 g/mL
1.600 to 1.700 g/mL
1.700 to 1.800 g/mL
Length (Metric):
250 mm
270 mm
300 mm
Unit Size:
Each
23 product options available for selection
| Product Code. | Specific Gravity Range | Length (Metric) | Unit Size |
|---|---|---|---|
| 11743193 | 1.300 to 1.400 g/mL | 300 mm | Each |
| 11763183 | 0.700 to 0.800 g/mL | 300 mm | Each |
| 11783183 | 0.900 to 1.000 g/mL | 300 mm | Each |
| 11753193 | 1.400 to 1.500 g/mL | 300 mm | Each |
| 11763193 | 1.500 to 1.600 g/mL | 300 mm | Each |
| 11773193 | 1.600 to 1.700 g/mL | 300 mm | Each |
| 11783193 | 1.700 to 1.800 g/mL | 300 mm | Each |
| 11783413 | 0.600 to 0.700 g/mL | 250 mm | Each |
| 11793413 | 0.700 to 0.800 g/mL | 250 mm | Each |
| 11703423 | 0.800 to 0.900 g/mL | 250 mm | Each |
| 11763423 | 0.140 to 0.150 g/mL | 250 mm | Each |
| 11793423 | 0.170 to 0.180 g/mL | 250 mm | Each |
| 11773423 | 0.150 to 0.160 g/mL | 250 mm | Each |
| 11783423 | 0.160 to 0.170 g/mL | 250 mm | Each |
| 11703433 | 0.180 to 0.190 g/mL | 250 mm | Each |
| 11773433 | 0.900 to 0.950 g/mL | 270 mm | Each |
| 11763433 | 0.850 to 0.900 g/mL | 270 mm | Each |
| 11743443 | 1.050 to 1.100 g/mL | 270 mm | Each |
| 11758083 | 1.200 to 1.250 g/mL | 270 mm | Each |
| 11768083 | 1.250 to 1.300 g/mL | 270 mm | Each |
| 11778083 | 1.300 to 1.350 g/mL | 270 mm | Each |
| 11788083 | 1.350 to 1.400 g/mL | 270 mm | Each |
| 11798083 | 1.400 to 1.450 g/mL | 270 mm | Each |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 23 options available.
Specifications
| Quantity | 1Each |
| Type | Density Hydrometer |
| Accuracy | ±0.002g/mL |
| Length (Metric) | 300 mm |
| For Use With (Application) | For general use |
| Specific Gravity Range | 1.300 to 1.400 g/mL |