Learn More
- Home
- Products
- Humidity and Hygrometry
- Hydrometers
- Alla France™ General Purpose Density Hydrometer
Alla France™ General Purpose Density Hydrometer
Product Code. 11783193
Change view
Click to view available options
Specific Gravity Range:
0.140 to 0.150 g/mL
0.150 to 0.160 g/mL
0.160 to 0.170 g/mL
0.170 to 0.180 g/mL
0.180 to 0.190 g/mL
0.600 to 0.700 g/mL
0.700 to 0.800 g/mL
0.800 to 0.900 g/mL
0.850 to 0.900 g/mL
0.900 to 0.950 g/mL
0.900 to 1.000 g/mL
1.050 to 1.100 g/mL
1.200 to 1.250 g/mL
1.250 to 1.300 g/mL
1.300 to 1.350 g/mL
1.300 to 1.400 g/mL
1.350 to 1.400 g/mL
1.400 to 1.450 g/mL
1.400 to 1.500 g/mL
1.500 to 1.600 g/mL
1.600 to 1.700 g/mL
1.700 to 1.800 g/mL
Length (Metric):
250 mm
270 mm
300 mm
Unit Size:
Each
23 product options available for selection
| Product Code. | Specific Gravity Range | Length (Metric) | Unit Size |
|---|---|---|---|
| 11783193 | 1.700 to 1.800 g/mL | 300 mm | Each |
| 11763183 | 0.700 to 0.800 g/mL | 300 mm | Each |
| 11783183 | 0.900 to 1.000 g/mL | 300 mm | Each |
| 11753193 | 1.400 to 1.500 g/mL | 300 mm | Each |
| 11743193 | 1.300 to 1.400 g/mL | 300 mm | Each |
| 11763193 | 1.500 to 1.600 g/mL | 300 mm | Each |
| 11773193 | 1.600 to 1.700 g/mL | 300 mm | Each |
| 11783413 | 0.600 to 0.700 g/mL | 250 mm | Each |
| 11793413 | 0.700 to 0.800 g/mL | 250 mm | Each |
| 11703423 | 0.800 to 0.900 g/mL | 250 mm | Each |
| 11763423 | 0.140 to 0.150 g/mL | 250 mm | Each |
| 11793423 | 0.170 to 0.180 g/mL | 250 mm | Each |
| 11773423 | 0.150 to 0.160 g/mL | 250 mm | Each |
| 11783423 | 0.160 to 0.170 g/mL | 250 mm | Each |
| 11703433 | 0.180 to 0.190 g/mL | 250 mm | Each |
| 11773433 | 0.900 to 0.950 g/mL | 270 mm | Each |
| 11763433 | 0.850 to 0.900 g/mL | 270 mm | Each |
| 11743443 | 1.050 to 1.100 g/mL | 270 mm | Each |
| 11758083 | 1.200 to 1.250 g/mL | 270 mm | Each |
| 11768083 | 1.250 to 1.300 g/mL | 270 mm | Each |
| 11778083 | 1.300 to 1.350 g/mL | 270 mm | Each |
| 11788083 | 1.350 to 1.400 g/mL | 270 mm | Each |
| 11798083 | 1.400 to 1.450 g/mL | 270 mm | Each |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 23 options available.
Product Code.
11783193
Supplier
Alla France™
Supplier No.
3010FG170/20qp
-
93.76€
36.91€ / Pack of 1
Save 56.85€
60% Off
You must Sign In and have a business account to purchase this item.
Restricted Item
In Stock
Log in to see stock.
Log in to see stock.
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Specifications
| Quantity | 1Each |
| Type | Density Hydrometer |
| Accuracy | ±0.002g/mL |
| Length (Metric) | 300 mm |
| For Use With (Application) | For general use |
| Specific Gravity Range | 1.700 to 1.800 g/mL |