missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SIRPB1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583866
This item is not returnable.
View return policy
Description
Immunogen sequence: VSKSYALEIS AHQKEHGSDI THEPALAPTA PL Highest antigen sequence indentity to the following orthologs: Mouse - 34%, Rat - 34%.
SIRP beta 1 is a type I transmembrane protein belonging to the SIRP family within the Ig superfamily. Members of this family are characterized by an extracellular region containing a V-set Ig domain with a J-like sequence and two C1-set Ig domains. Unlike SIRP alpha that has cytoplasmic ITIM domains, SIRP beta 1 possesses positively charged residues that allow association with ITAM motif-containing adaptor molecules. SIRP beta 1 is expressed on cells of monocyte, macrophage or dendritic lineages.
Specifications
| SIRPB1 | |
| Polyclonal | |
| Unconjugated | |
| SIRPB1 | |
| 9930027N05Rik; CD172 antigen-like family member B; CD172b; CD172b antigen; DKFZp686A05192; signal regulatory protein beta 1; signal-regulatory protein beta 1; signal-regulatory protein beta 1A; signal-regulatory protein beta-1; Signal-regulatory protein beta-1 isoform 3; SIRP BETA 1; SIRP beta 1 cell surface protein; Sirpb; SIRPB1; Sirpb1a; SIRP-beta; SIRP-beta B; SIRP-BETA-1; SIRP-beta-1 isoform 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10326 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O00241 | |
| SIRPB1 | |
| Recombinant protein corresponding to Human SIRPB1. Recombinant protein control fragment (Product #RP-100868). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction