missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Gephyrin Monoclonal Antibody (24GB5580)

Mouse Monoclonal Antibody
198.00€ - 564.00€
Specifications
| Antigen | Gephyrin |
|---|---|
| Clone | 24GB5580 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30276258
![]() |
Invitrogen™
MA557410 |
100 μL |
564.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30276232
|
Invitrogen™
MA557411 |
20 μL |
198.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Gephyrin Monoclonal Antibody for Western Blot, Flow
Gephyrin is a neuronal assembly protein that anchors inhibitory neurotransmitter receptors to the postsynaptic cytoskeleton via high affinity binding to a receptor subunit domain and tubulin dimers. In nonneuronal tissues, the encoded protein is also required for molybdenum cofactor biosynthesis. Mutations in this gene may be associated with the neurological condition hyperplexia and also lead to molybdenum cofactor deficiency. Numerous alternatively spliced transcript variants encoding different isoforms have been described; however, the full-length nature of all transcript variants is not currently known.Specifications
| Gephyrin | |
| -20°C | |
| Monoclonal | |
| Liquid | |
| IgG2a | |
| Human, Mouse | |
| Gphn | |
| 5730552E08Rik; AI662856; BC027112; C230040D23; Domain E; Domain G; EGK_18299; Geph; gephyrin; gephyrin isoform; Gph; GPHN; GPHRYN; HKPX1; KIAA1385; MOCODC; Molybdopterin adenylyltransferase; Molybdopterin molybdenumtransferase; MPT adenylyltransferase; MPT Mo-transferase; Putative glycine receptor-tubulin linker protein | |
| Gphn | |
| Primary | |
| Affinity chromatography |
| 24GB5580 | |
| Flow Cytometry, Western Blot | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8BUV3, Q9NQX3 | |
| 10243, 268566 | |
| Recombinant protein of human GPHN (aa 1-736). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title