missing translation for 'onlineSavingsMsg'
Learn More

GFAP Antibody (rGFAP/9150), HRP, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-24050H

579.70 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.


Product Code. 18650234

  • 679.00€ / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

GFAP Monoclonal antibody specifically detects GFAP in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GFAP
Monoclonal
HRP
ALXDRD, FLJ45472, GFAP, GFAP astrocytes, glial fibrillary acidic protein
Recombinant fragment (around aa100-300) of human GFAP protein (exact sequence is proprietary)
0.1 mL
Astrocyte Markers, Cancer, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Inflammation, Neurodegeneration, Neuroscience, Signal Transduction, Stem Cell Markers
2670
Store at 4°C in the dark.
IgG2b κ
Immunohistochemistry (Paraffin)
rGFAP/9150
PBS
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.