missing translation for 'onlineSavingsMsg'
Learn More

GFAP Mouse anti-Human, Mouse, Porcine, Rat, Clone: 4B2E10, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 60190-1-IG-150UL

 View more versions of this product

Product Code. 16877175

  • 410.00€ / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

GFAP (Glial fibrillary acidic protein) is a member of the class III intermediate filament protein family. GFAP is heavily, and specifically, expressed in astrocytes and certain astroglia in the central nervous system, in satellite cells in peripheral ganglia, and in non-myelinating Schwann cells in peripheral nerves. In addition, neural stem cells strongly express GFAP. Antibodies to GFAP are very useful as markers of astrocytic cells. In addition, many types of brain tumor, presumably derived from astrocytic cells, heavily express GFAP. GFAP is also found in the lens epithelium, Kupffer cells of the liver, in some cells in salivary tumors and has been reported in erythrocytes. GFAP is used as a marker to distinguish astrocytes from other glial cells during development. Mutations in this gene cause Alexander disease, a rare disorder of astrocytes in the central nervous system. Alternative splicing of the GFAP gene results in multiple transcript variants encoding distinct isoforms.
Specifications

Specifications

GFAP
Monoclonal
1.5 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P03995, P14136, P47819
GFAP
GFAP Fusion Protein Ag10452
150 μL
Primary
Human, Mouse, Pig, Rat
Antibody
IgG2a
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
4B2E10
Unconjugated
GFAP
GFAP
Mouse
Protein A
RUO
14580, 24387, 2670, 396562
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.