missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFAP Rabbit anti-Human, Mouse, Pig, Rat, Clone: 4C6, Proteintech
Rabbit Recombinant Monoclonal Antibody
149.00€ - 439.00€
Specifications
| Antigen | GFAP |
|---|---|
| Clone | 4C6 |
| Concentration | 1000 μg/mL |
| Applications | Immunohistochemistry, Western Blot, ELISA |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17721219
|
Proteintech
81063-1-RR-20UL |
20 μL |
149.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17711219
|
Proteintech
81063-1-RR-100UL |
100 μL |
439.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GFAP Recombinant Monoclonal antibody specifically detects GFAP in Human, Mouse, Pig, Rat samples. It is validated for IHC, WB, ELISASpecifications
| GFAP | |
| 1000 μg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| Human, Mouse, Pig, Rat | |
| 2670 | |
| Primary | |
| Store at -20°C |
| 4C6 | |
| Immunohistochemistry, Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
| IgG | |
| Protein A purification | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title