Learn More
GFPT1 Antibody (1F1A4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52472
 View more versions of this product
Product Code. 18260381
  • 552.00€ / 0.10mg
Description
GFPT1 Monoclonal specifically detects GFPT1 in Human, Rat, Monkey samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GFPT1
Monoclonal
1 mg/mL
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, CyTOF-ready
glucosamine--fructose-6-phosphate aminotransferase [isomerizing] 1, CMSTA1, GFA, GFAT, GFAT 1, GFAT1, GFAT1m, GFPT, GFPT1L, glutamine--fructose-6-phosphate transaminase 1, MSLG
Mouse
78.8 kDa
0.1 mg
metabolism
2673
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG1
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, CyTOF
1F1A4
Unconjugated
PBS with 0.05% Sodium Azide
GFPT1
Purified recombinant fragment of human GFPT1 (AA: 536-681) expressed in E. Coli.
Protein A or G purified
RUO
Primary
Human, Rat, Monkey